Product Details

SNP ID
rs201081455
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67405120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTGCAGACCTCATCCGCCAAC[A/G]GGAGATGAAGTGGGTGGAGATGACC
Phenotype
MIM: 176875 MIM: 603761 MIM: 610831
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PPP1CA PubMed Links

Gene Details

Gene
PPP1CA
Gene Name
protein phosphatase 1 catalytic subunit alpha
There are no transcripts associated with this gene.

Gene
RAD9A
Gene Name
RAD9 checkpoint clamp component A
There are no transcripts associated with this gene.

Gene
TBC1D10C
Gene Name
TBC1 domain family member 10C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256508.1 504 Missense Mutation CAG,CGG Q63R NP_001243437.1
NM_198517.3 504 Missense Mutation CAG,CGG Q63R NP_940919.1
XM_006718538.3 504 Missense Mutation CAG,CGG Q63R XP_006718601.1
XM_006718539.3 504 Missense Mutation CAG,CGG Q63R XP_006718602.1
XM_006718540.3 504 Missense Mutation CAG,CGG Q63R XP_006718603.1
XM_006718541.3 504 UTR 5 XP_006718604.1
XM_006718542.3 504 UTR 5 XP_006718605.1
XM_006718543.2 504 UTR 5 XP_006718606.1
XM_006718544.3 504 UTR 5 XP_006718607.1
XM_011545001.2 504 UTR 5 XP_011543303.1
XM_011545002.2 504 Missense Mutation CAG,CGG Q63R XP_011543304.1

View Full Product Details