Product Details

SNP ID
rs200508281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27082463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCTGTCGTCCTCTATGTCGCT[C/G]TTAGAGGATGAATATGCCATGGAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C12orf71 PubMed Links

Gene Details

Gene
C12orf71
Gene Name
chromosome 12 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080406.1 602 Missense Mutation AAC,AAG N7K NP_001073875.1
XM_011520847.2 602 Missense Mutation AAC,AAG N7K XP_011519149.1

View Full Product Details