Product Details

SNP ID
rs201634299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52691331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCGTGCCCCGCCTCCGCGGTAG[C/T]TTCTTCCGCCGCCATAGCCCCCACC
Phenotype
MIM: 611158
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT77 PubMed Links

Gene Details

Gene
KRT77
Gene Name
keratin 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175078.2 1362 Missense Mutation NP_778253.2
XM_011538288.2 1362 Missense Mutation XP_011536590.1
XM_011538289.2 1362 Intron XP_011536591.1

View Full Product Details