Product Details

SNP ID
rs202207799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52362279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCTCGCCCTCCAGCAGGCGCC[C/T]GTAGGTGGCGATCTCGATGTCCAGG
Phenotype
MIM: 602767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT85 PubMed Links

Gene Details

Gene
KRT85
Gene Name
keratin 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300810.1 836 Missense Mutation AGG,GGG R212G NP_001287739.1
NM_002283.3 836 Missense Mutation AGG,GGG R424G NP_002274.1

View Full Product Details