Product Details

SNP ID
rs201284757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54292990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTACATCTCTACATATTCGCTG[C/T]GCCGCCGACCAGCCTCTGTCCCCTC
Phenotype
MIM: 164017 MIM: 601490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPA1 PubMed Links

Gene Details

Gene
HNRNPA1
Gene Name
heterogeneous nuclear ribonucleoprotein A1
There are no transcripts associated with this gene.

Gene
NFE2
Gene Name
nuclear factor, erythroid 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136023.2 929 Missense Mutation CAC,CGC H169R NP_001129495.1
NM_001261461.1 929 Missense Mutation CAC,CGC H169R NP_001248390.1
NM_006163.2 929 Missense Mutation CAC,CGC H169R NP_006154.1
XM_005268906.4 929 Missense Mutation CAC,CGC H169R XP_005268963.1

View Full Product Details