Product Details

SNP ID
rs201271144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60713500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGTCTTCACAGACAGTATCAAC[C/T]ATGCCATCTAAGACATACTTGGATT
Phenotype
MIM: 606342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIX4 PubMed Links

Gene Details

Gene
SIX4
Gene Name
SIX homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017420.4 2313 Missense Mutation ATA,ATG I751M NP_059116.3
XM_005267759.2 2313 Missense Mutation ATA,ATG I743M XP_005267816.1

View Full Product Details