Product Details

SNP ID
rs200132448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43329476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTACGCTGGCAGGGAATACCCC[C/T]GAAGGCGAAGCAGGATTTACGCGAG
Phenotype
MIM: 611246
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAL PubMed Links

Gene Details

Gene
ADAL
Gene Name
adenosine deaminase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012969.3 1147 Intron NP_001012987.1
NM_001159280.2 1147 Intron NP_001152752.1
NM_001324364.1 1147 Intron NP_001311293.1
NM_001324365.1 1147 Intron NP_001311294.1
NM_001324366.1 1147 Intron NP_001311295.1
NM_001324367.1 1147 Intron NP_001311296.1
NM_001324368.1 1147 Intron NP_001311297.1
XM_011521291.2 1147 Intron XP_011519593.1
XM_017021964.1 1147 Intron XP_016877453.1
XM_017021965.1 1147 Intron XP_016877454.1
XM_017021966.1 1147 Intron XP_016877455.1
XM_017021967.1 1147 Intron XP_016877456.1
XM_017021968.1 1147 Intron XP_016877457.1
XM_017021969.1 1147 Intron XP_016877458.1
Gene
LCMT2
Gene Name
leucine carboxyl methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014793.4 1147 Silent Mutation TCA,TCG S338S NP_055608.2

View Full Product Details