Product Details

SNP ID
rs199667793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66505367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGCGGCTGAACTGCGCAGCAG[A/T]GGACTTTTATTCTCGTCTCCTTCAG
Phenotype
MIM: 180479 MIM: 605979 MIM: 609984
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR4512 PubMed Links

Gene Details

Gene
MIR4512
Gene Name
microRNA 4512
There are no transcripts associated with this gene.

Gene
RPL4
Gene Name
ribosomal protein L4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000968.3 285 Intron NP_000959.2
Gene
SNAPC5
Gene Name
small nuclear RNA activating complex polypeptide 5
There are no transcripts associated with this gene.

Gene
SNORD16
Gene Name
small nucleolar RNA, C/D box 16
There are no transcripts associated with this gene.

Gene
SNORD18A
Gene Name
small nucleolar RNA, C/D box 18A
There are no transcripts associated with this gene.

Gene
SNORD18B
Gene Name
small nucleolar RNA, C/D box 18B
There are no transcripts associated with this gene.

Gene
SNORD18C
Gene Name
small nucleolar RNA, C/D box 18C
There are no transcripts associated with this gene.

Gene
ZWILCH
Gene Name
zwilch kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287821.1 285 UTR 5 NP_001274750.1
NM_001287822.1 285 UTR 5 NP_001274751.1
NM_001287823.1 285 UTR 5 NP_001274752.1
NM_017975.4 285 Missense Mutation GAG,GTG E10V NP_060445.3

View Full Product Details