Product Details

SNP ID
rs200402711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:23566264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGGAAGTGGCGGAAGCCCCCC[C/T]GGCTGCATCCTCCCTTTCCTTGCCT
Phenotype
MIM: 603856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4508 PubMed Links

Gene Details

Gene
MIR4508
Gene Name
microRNA 4508
There are no transcripts associated with this gene.

Gene
MKRN3
Gene Name
makorin ring finger protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005664.3 958 Missense Mutation CCG,CTG P161L NP_005655.1

View Full Product Details