Product Details

SNP ID
rs201110852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29744777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGGGGTCTGGGCGGAGAGCAC[A/G]GCGGAACAGGCGTTCAGCCAACACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf54 PubMed Links

Gene Details

Gene
C16orf54
Gene Name
chromosome 16 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175900.3 284 Missense Mutation CGT,TGT R59C NP_787096.2

View Full Product Details