Product Details

SNP ID
rs199711526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67940149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCTCATAGAGCACACCCACAG[G/T]GTCCGTGTAGGGGAAGCCGTGGTCG
Phenotype
MIM: 118888 MIM: 606967 MIM: 176847 MIM: 604119
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CTRL PubMed Links

Gene Details

Gene
CTRL
Gene Name
chymotrypsin like
There are no transcripts associated with this gene.

Gene
LCAT
Gene Name
lecithin-cholesterol acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000229.1 1089 Missense Mutation ACT,CCT T360P NP_000220.1
Gene
PSMB10
Gene Name
proteasome subunit beta 10
There are no transcripts associated with this gene.

Gene
SLC12A4
Gene Name
solute carrier family 12 member 4
There are no transcripts associated with this gene.

View Full Product Details