Product Details

SNP ID
rs200936245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1446679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAAGCCTCTCTTTCCCAGGCG[A/G]TACCTGGCGAGGTCCTTCCTGGTCA
Phenotype
MIM: 602727
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC154 PubMed Links

Gene Details

Gene
CCDC154
Gene Name
coiled-coil domain containing 154
There are no transcripts associated with this gene.

Gene
CLCN7
Gene Name
chloride voltage-gated channel 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114331.2 2408 Silent Mutation TAC,TAT Y766Y NP_001107803.1
NM_001287.5 2408 Silent Mutation TAC,TAT Y790Y NP_001278.1
XM_011522354.1 2408 Silent Mutation TAC,TAT Y732Y XP_011520656.1

View Full Product Details