Product Details

SNP ID
rs201361839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1446687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCTTTCCCAGGCGGTACCTGG[C/T]GAGGTCCTTCCTGGTCACCAACCCG
Phenotype
MIM: 602727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC154 PubMed Links

Gene Details

Gene
CCDC154
Gene Name
coiled-coil domain containing 154
There are no transcripts associated with this gene.

Gene
CLCN7
Gene Name
chloride voltage-gated channel 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114331.2 2400 Missense Mutation ACC,GCC T764A NP_001107803.1
NM_001287.5 2400 Missense Mutation ACC,GCC T788A NP_001278.1
XM_011522354.1 2400 Missense Mutation ACC,GCC T730A XP_011520656.1

View Full Product Details