Product Details

SNP ID
rs202070785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3494566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGGGCAATCGGGCTTCCAGAA[C/G]GGCTGAACAGAGGCTGTTACGTGGG
Phenotype
MIM: 616787 MIM: 614246
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C16orf90 PubMed Links

Gene Details

Gene
C16orf90
Gene Name
chromosome 16 open reading frame 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080524.1 424 Missense Mutation CTT,GTT L120V NP_001073993.1
XM_005255502.3 424 Missense Mutation CTT,GTT L135V XP_005255559.1
Gene
CLUAP1
Gene Name
clusterin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015041.2 424 Intron NP_055856.1
NM_024793.2 424 Intron NP_079069.1
XM_006720867.3 424 Intron XP_006720930.1
XM_017023068.1 424 Intron XP_016878557.1
XM_017023069.1 424 Intron XP_016878558.1
XM_017023070.1 424 Intron XP_016878559.1
XM_017023071.1 424 Intron XP_016878560.1
XM_017023072.1 424 Intron XP_016878561.1
XM_017023073.1 424 Intron XP_016878562.1
Gene
MIR6126
Gene Name
microRNA 6126
There are no transcripts associated with this gene.

Gene
NAA60
Gene Name
N(alpha)-acetyltransferase 60, NatF catalytic subunit
There are no transcripts associated with this gene.

View Full Product Details