Product Details

SNP ID
rs199988811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45024438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGCCTTGAGTGCCCCACTAGG[C/T]GTAAGGCAGAAGGTAGTGGGTGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAKD PubMed Links

Gene Details

Gene
DCAKD
Gene Name
dephospho-CoA kinase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128631.2 1047 Missense Mutation ACC,GCC T231A NP_001122103.1
NM_001288654.2 1047 Missense Mutation ACC,GCC T231A NP_001275583.1
NM_001288655.1 1047 Missense Mutation ACC,GCC T231A NP_001275584.1
NM_001321326.1 1047 Missense Mutation ACC,GCC T231A NP_001308255.1
NM_024819.6 1047 Missense Mutation ACC,GCC T231A NP_079095.3
XM_005257688.2 1047 Intron XP_005257745.1
XM_011525262.2 1047 Missense Mutation ACC,GCC T231A XP_011523564.1
XM_017025102.1 1047 Intron XP_016880591.1
XM_017025103.1 1047 Intron XP_016880592.1
XM_017025104.1 1047 Intron XP_016880593.1
XM_017025105.1 1047 Missense Mutation ACC,GCC T130A XP_016880594.1
XM_017025106.1 1047 Missense Mutation ACC,GCC T130A XP_016880595.1

View Full Product Details