Product Details

SNP ID
rs200058537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15987298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACGCCTTCCACTGGGTGATGAGA[C/T]TAAGGTGATGGACTGTCGATCAACT
Phenotype
MIM: 614535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSWIM7 PubMed Links

Gene Details

Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 267 Missense Mutation ATC,GTC I57V NP_001036162.1
NM_001042698.1 267 Missense Mutation ATC,GTC I57V NP_001036163.1

View Full Product Details