Product Details

SNP ID
rs202059159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48051333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTATGGTATCCACCCCAAGAGCA[C/T]AGACCTGGACAATTACTTCACTGCC
Phenotype
MIM: 615526 MIM: 163260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPZ2 PubMed Links

Gene Details

Gene
COPZ2
Gene Name
coatomer protein complex subunit zeta 2
There are no transcripts associated with this gene.

Gene
NFE2L1
Gene Name
nuclear factor, erythroid 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003204.2 866 Missense Mutation ACA,ATA T72I NP_003195.1
XM_005257410.3 866 Missense Mutation ACA,ATA T72I XP_005257467.1
XM_005257411.3 866 Missense Mutation ACA,ATA T72I XP_005257468.1
XM_005257412.3 866 Missense Mutation ACA,ATA T72I XP_005257469.1
XM_005257413.3 866 Missense Mutation ACA,ATA T72I XP_005257470.1
XM_005257415.3 866 Missense Mutation ACA,ATA T72I XP_005257472.1
XM_017024691.1 866 Missense Mutation ACA,ATA T72I XP_016880180.1

View Full Product Details