Product Details
- SNP ID
-
rs201038576
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:62565081 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCCACTTTACTACTGTCCGACA[C/T]GGAGCCTCATTTACTGAACAGTGGA
- Phenotype
-
MIM: 608439
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLK2
PubMed Links
Gene Details
- Gene
- TLK2
- Gene Name
- tousled like kinase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001284333.1 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
NP_001271262.1 |
NM_001284363.1 |
1017 |
Intron |
|
|
NP_001271292.1 |
NM_006852.3 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
NP_006843.2 |
XM_011524215.2 |
1017 |
Silent Mutation |
CAC,CAT |
H273H |
XP_011522517.1 |
XM_011524216.2 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
XP_011522518.1 |
XM_011524217.2 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
XP_011522519.1 |
XM_011524218.2 |
1017 |
Silent Mutation |
CAC,CAT |
H273H |
XP_011522520.1 |
XM_011524220.2 |
1017 |
Silent Mutation |
CAC,CAT |
H241H |
XP_011522522.1 |
XM_011524221.2 |
1017 |
Silent Mutation |
CAC,CAT |
H272H |
XP_011522523.1 |
XM_011524222.2 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
XP_011522524.1 |
XM_011524223.2 |
1017 |
Silent Mutation |
CAC,CAT |
H241H |
XP_011522525.1 |
XM_011524224.2 |
1017 |
Silent Mutation |
CAC,CAT |
H200H |
XP_011522526.1 |
XM_011524225.2 |
1017 |
Silent Mutation |
CAC,CAT |
H200H |
XP_011522527.1 |
XM_011524226.2 |
1017 |
Silent Mutation |
CAC,CAT |
H200H |
XP_011522528.1 |
XM_011524227.2 |
1017 |
Silent Mutation |
CAC,CAT |
H200H |
XP_011522529.1 |
XM_011524228.2 |
1017 |
Silent Mutation |
CAC,CAT |
H200H |
XP_011522530.1 |
XM_011524229.2 |
1017 |
Silent Mutation |
CAC,CAT |
H155H |
XP_011522531.1 |
XM_011524231.1 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
XP_011522533.1 |
XM_017024044.1 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
XP_016879533.1 |
XM_017024045.1 |
1017 |
Silent Mutation |
CAC,CAT |
H318H |
XP_016879534.1 |
XM_017024046.1 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
XP_016879535.1 |
XM_017024047.1 |
1017 |
Silent Mutation |
CAC,CAT |
H304H |
XP_016879536.1 |
XM_017024048.1 |
1017 |
Silent Mutation |
CAC,CAT |
H272H |
XP_016879537.1 |
XM_017024049.1 |
1017 |
Silent Mutation |
CAC,CAT |
H272H |
XP_016879538.1 |
XM_017024050.1 |
1017 |
Silent Mutation |
CAC,CAT |
H287H |
XP_016879539.1 |
XM_017024051.1 |
1017 |
Silent Mutation |
CAC,CAT |
H272H |
XP_016879540.1 |
XM_017024052.1 |
1017 |
Silent Mutation |
CAC,CAT |
H272H |
XP_016879541.1 |
XM_017024053.1 |
1017 |
Silent Mutation |
CAC,CAT |
H272H |
XP_016879542.1 |
XM_017024054.1 |
1017 |
Silent Mutation |
CAC,CAT |
H241H |
XP_016879543.1 |
XM_017024055.1 |
1017 |
Silent Mutation |
CAC,CAT |
H209H |
XP_016879544.1 |
XM_017024056.1 |
1017 |
Silent Mutation |
CAC,CAT |
H200H |
XP_016879545.1 |
XM_017024057.1 |
1017 |
Silent Mutation |
CAC,CAT |
H200H |
XP_016879546.1 |
XM_017024058.1 |
1017 |
Silent Mutation |
CAC,CAT |
H155H |
XP_016879547.1 |
XM_017024059.1 |
1017 |
Silent Mutation |
CAC,CAT |
H155H |
XP_016879548.1 |
XM_017024060.1 |
1017 |
Silent Mutation |
CAC,CAT |
H92H |
XP_016879549.1 |
View Full Product Details