Product Details

SNP ID
rs201210478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50064097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGCTCCTGGCTGGTGCCCCCC[A/G]GGAGCTCGCTGTGCCCGATGGCTAC
Phenotype
MIM: 605025
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGA3 PubMed Links

Gene Details

Gene
ITGA3
Gene Name
integrin subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002204.3 602 Missense Mutation CAG,CGG Q76R NP_002195.1
XM_005257308.2 602 Missense Mutation CAG,CGG Q76R XP_005257365.1
Gene
LOC284080
Gene Name
uncharacterized LOC284080
There are no transcripts associated with this gene.

View Full Product Details