Product Details

SNP ID
rs201932039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3215884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCTGGCCAACCATCTCTTGGG[C/G]AGCAAATCCATCTCTTTTGGGGGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR1A1 PubMed Links

Gene Details

Gene
OR1A1
Gene Name
olfactory receptor family 1 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014565.2 264 Silent Mutation GGC,GGG G88G NP_055380.2

View Full Product Details