Product Details

SNP ID
rs201456706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82657936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGGGCGGACGAGCTTCACTTTG[C/T]GGAGGCTGCTCCTTTTGTGGGTGGA
Phenotype
MIM: 609226
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB40B PubMed Links

Gene Details

Gene
RAB40B
Gene Name
RAB40B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006822.2 1002 Missense Mutation CAC,CGC H255R NP_006813.1
XM_005256334.4 1002 Missense Mutation CAC,CGC H195R XP_005256391.1
XM_005256335.3 1002 Intron XP_005256392.1
XM_006722271.3 1002 Missense Mutation CAC,CGC H215R XP_006722334.1
XM_011523528.2 1002 Missense Mutation CAC,CGC H238R XP_011521830.1
XM_017024042.1 1002 Intron XP_016879531.1
XM_017024043.1 1002 Missense Mutation CAC,CGC H195R XP_016879532.1
Gene
WDR45B
Gene Name
WD repeat domain 45B
There are no transcripts associated with this gene.

View Full Product Details