Product Details

SNP ID
rs201256045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4960033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCATAGCTCACCTTCAGCTTCC[A/G]CTTCTCCTCAGCCTTCTGGGGGTCC
Phenotype
MIM: 611508 MIM: 131370 MIM: 610056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMTA2 PubMed Links

Gene Details

Gene
CAMTA2
Gene Name
calmodulin binding transcription activator 2
There are no transcripts associated with this gene.

Gene
ENO3
Gene Name
enolase 3
There are no transcripts associated with this gene.

Gene
MIR6864
Gene Name
microRNA 6864
There are no transcripts associated with this gene.

Gene
SPAG7
Gene Name
sperm associated antigen 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004890.2 1064 Missense Mutation CGG,TGG R136W NP_004881.2
XM_006721600.2 1064 Missense Mutation CGG,TGG R85W XP_006721663.1

View Full Product Details