Product Details

SNP ID
rs200009297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50283282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTTACTTTGGGGTCCCGTGAGT[A/G]CTCGGGGCACAGCACCTGGAGCCGC
Phenotype
MIM: 609150 MIM: 156535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXXC1 PubMed Links

Gene Details

Gene
CXXC1
Gene Name
CXXC finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101654.1 2132 Missense Mutation CAC,TAC H556Y NP_001095124.1
NM_014593.3 2132 Missense Mutation CAC,TAC H552Y NP_055408.2
XM_011525940.2 2132 Missense Mutation CAC,TAC H556Y XP_011524242.1
XM_011525941.2 2132 Missense Mutation CAC,TAC H431Y XP_011524243.1
XM_017025718.1 2132 Missense Mutation CAC,TAC H552Y XP_016881207.1
XM_017025719.1 2132 Missense Mutation CAC,TAC H427Y XP_016881208.1
Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
There are no transcripts associated with this gene.

View Full Product Details