Product Details

SNP ID
rs200134520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57720633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGTCTTTGAGTGAAGGCTTTCC[C/T]GCACTTGCTGCACTCATAAGGCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF671 PubMed Links

Gene Details

Gene
ZNF671
Gene Name
zinc finger protein 671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321375.1 2045 Missense Mutation AGG,GGG R387G NP_001308304.1
NM_001321376.1 2045 Missense Mutation AGG,GGG R408G NP_001308305.1
NM_024833.2 2045 Missense Mutation AGG,GGG R485G NP_079109.2
XM_017027314.1 2045 Missense Mutation AGG,GGG R408G XP_016882803.1

View Full Product Details