Product Details
- SNP ID
-
rs202168345
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55433694 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCGCCCCAGGCCTCCATGCGATAG[A/C]CGCCCCCCGTGCCCGGCCGCCGCAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA7
PubMed Links
Gene Details
- Gene
- SHISA7
- Gene Name
- shisa family member 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145176.1 |
1079 |
Missense Mutation |
GGC,GTC |
G360V |
NP_001138648.1 |
View Full Product Details