Product Details

SNP ID
rs200748250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36891602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCAGTGTGAATTCTCTGATGTC[A/G]AGTAAGATTTGAGCCTTTATTAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF345 PubMed Links

Gene Details

Gene
ZNF345
Gene Name
zinc finger protein 345
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242472.1 1554 Intron NP_001229401.1
NM_001242474.1 1554 Intron NP_001229403.1
NM_001242475.1 1554 Intron NP_001229404.1
NM_001242476.1 1554 Intron NP_001229405.1
NM_003419.4 1554 Intron NP_003410.1
XM_011526704.2 1554 Intron XP_011525006.1
XM_017026568.1 1554 Intron XP_016882057.1
XM_017026569.1 1554 Intron XP_016882058.1
XM_017026570.1 1554 Intron XP_016882059.1
XM_017026571.1 1554 Intron XP_016882060.1
XM_017026572.1 1554 Intron XP_016882061.1
XM_017026573.1 1554 Intron XP_016882062.1
Gene
ZNF829
Gene Name
zinc finger protein 829
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037232.3 1554 Nonsense Mutation CGA,TGA R397* NP_001032309.2
NM_001171979.1 1554 Nonsense Mutation CGA,TGA R478* NP_001165450.1
XM_005258876.4 1554 Nonsense Mutation CGA,TGA R397* XP_005258933.1
XM_011526933.2 1554 Nonsense Mutation CGA,TGA R397* XP_011525235.1

View Full Product Details