Product Details

SNP ID
rs201095571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57720523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAACCCTCTGGTGCAGAACAAG[G/T]GTCTGTTTCCGGATGAATTCTCTCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF671 PubMed Links

Gene Details

Gene
ZNF671
Gene Name
zinc finger protein 671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321375.1 2155 Silent Mutation ACA,ACC T423T NP_001308304.1
NM_001321376.1 2155 Silent Mutation ACA,ACC T444T NP_001308305.1
NM_024833.2 2155 Silent Mutation ACA,ACC T521T NP_079109.2
XM_017027314.1 2155 Silent Mutation ACA,ACC T444T XP_016882803.1

View Full Product Details