Product Details

SNP ID
rs201753059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57720634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTCTTTGAGTGAAGGCTTTCCC[A/G]CACTTGCTGCACTCATAAGGCTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF671 PubMed Links

Gene Details

Gene
ZNF671
Gene Name
zinc finger protein 671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321375.1 2044 Silent Mutation TGC,TGT C386C NP_001308304.1
NM_001321376.1 2044 Silent Mutation TGC,TGT C407C NP_001308305.1
NM_024833.2 2044 Silent Mutation TGC,TGT C484C NP_079109.2
XM_017027314.1 2044 Silent Mutation TGC,TGT C407C XP_016882803.1

View Full Product Details