Product Details

SNP ID
rs201791412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43767627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGTCAGGGCATCCAGCTTCCC[C/T]GCCAGCGTGTCAATCTGTTTCTCCA
Phenotype
MIM: 602754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNN4 PubMed Links

Gene Details

Gene
KCNN4
Gene Name
potassium calcium-activated channel subfamily N member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002250.2 1596 Silent Mutation GCA,GCG A400A NP_002241.1
XM_005258882.2 1596 Silent Mutation GCA,GCG A368A XP_005258939.1
XM_005258883.2 1596 Silent Mutation GCA,GCG A337A XP_005258940.1
XM_011526938.1 1596 Intron XP_011525240.1

View Full Product Details