Product Details

SNP ID
rs200998348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:853031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGGCCGCGCACTGCGTGGCGAAT[A/G]TGTGAGTAGCCGGGAGTGTGCGCGC
Phenotype
MIM: 134350 MIM: 130130 MIM: 177020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFD PubMed Links

Gene Details

Gene
CFD
Gene Name
complement factor D
There are no transcripts associated with this gene.

Gene
ELANE
Gene Name
elastase, neutrophil expressed
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001972.3 359 Missense Mutation ATA,GTA I75V NP_001963.1
XM_011527775.1 359 Missense Mutation ATA,GTA I75V XP_011526077.1
XM_011527776.1 359 Missense Mutation ATA,GTA I75V XP_011526078.1
Gene
PRTN3
Gene Name
proteinase 3
There are no transcripts associated with this gene.

View Full Product Details