Product Details

SNP ID
rs200727342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2102225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTCTAACAAGTTGCTCAGTAGC[A/G]TGGAGTCACTGCACTTCCCGTCGAC
Phenotype
MIM: 607246
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP3D1 PubMed Links

Gene Details

Gene
AP3D1
Gene Name
adaptor related protein complex 3 delta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261826.1 3116 Missense Mutation ACG,ATG T1199M NP_001248755.1
NM_003938.6 3116 Missense Mutation ACG,ATG T1137M NP_003929.4
XM_006722932.2 3116 Missense Mutation ACG,ATG T1187M XP_006722995.1
XM_017027422.1 3116 Missense Mutation ACG,ATG T971M XP_016882911.1
Gene
IZUMO4
Gene Name
IZUMO family member 4
There are no transcripts associated with this gene.

Gene
MOB3A
Gene Name
MOB kinase activator 3A
There are no transcripts associated with this gene.

View Full Product Details