Product Details

SNP ID
rs201107400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1555357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATACTGTCGTTGAAGGGCTGAGG[C/G]GCCGCCGGGCTGCGGGGTCTCCGTC
Phenotype
MIM: 611009
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MEX3D PubMed Links

Gene Details

Gene
MEX3D
Gene Name
mex-3 RNA binding family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174118.1 1975 Missense Mutation CCC,GCC P659A NP_001167589.1
NM_203304.3 1975 UTR 3 NP_976049.3
XM_017026811.1 1975 UTR 3 XP_016882300.1

View Full Product Details