Product Details

SNP ID
rs201283217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1556744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCTGGGCGGCGCCGGGCAGAC[A/C]CCCGGCCTTGCTGCGCGTGGCGCGG
Phenotype
MIM: 611009
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MEX3D PubMed Links

Gene Details

Gene
MEX3D
Gene Name
mex-3 RNA binding family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174118.1 775 Missense Mutation GGT,TGT G259C NP_001167589.1
NM_203304.3 775 Missense Mutation GGT,TGT G259C NP_976049.3
XM_017026811.1 775 Missense Mutation GGT,TGT G25C XP_016882300.1

View Full Product Details