Product Details

SNP ID
rs201324340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54594914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTACGGTAGCCACACTGCAGGCT[A/G]GTCAGAGCCCAGTGACCCCCTGGAG
Phenotype
MIM: 604810 MIM: 604812
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LILRA1 PubMed Links

Gene Details

Gene
LILRA1
Gene Name
leukocyte immunoglobulin like receptor A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278318.1 368 Intron NP_001265247.1
NM_001278319.1 368 Nonsense Mutation TAG,TGG *107W NP_001265248.1
NM_006863.3 368 Intron NP_006854.1
Gene
LILRA2
Gene Name
leukocyte immunoglobulin like receptor A2
There are no transcripts associated with this gene.

View Full Product Details