Product Details

SNP ID
rs2230350
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:109548825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAAAAGCGGCCAAAGAAGTGAA[A/G]CTGCTGCTACTCGGTGAGGGGCTGG
Phenotype
MIM: 139370 MIM: 606916
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAI3 PubMed Links

Gene Details

Gene
GNAI3
Gene Name
G protein subunit alpha i3
There are no transcripts associated with this gene.

Gene
GPR61
Gene Name
G protein-coupled receptor 61
There are no transcripts associated with this gene.

View Full Product Details