Product Details

SNP ID
rs201652917
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:74236130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGAAGAAAAAAGTCAGTGAATC[A/G]TATGTTATCACAATAGAAAGATTAG
Phenotype
MIM: 613932
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FPGT-TNNI3K PubMed Links

Gene Details

Gene
FPGT-TNNI3K
Gene Name
FPGT-TNNI3K readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112808.2 134 Silent Mutation TCA,TCG S137S NP_001106279.2
NM_001199327.1 134 Silent Mutation TCA,TCG S137S NP_001186256.2
Gene
TNNI3K
Gene Name
TNNI3 interacting kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015978.2 134 Silent Mutation TCA,TCG S23S NP_057062.1

View Full Product Details