Product Details

SNP ID
rs200980850
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:206637695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCTTCATGATGATAGATGAAAC[C/G]AAATGTCCCCCCTGTTCAAATGTAC
Phenotype
MIM: 603497
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DYRK3 PubMed Links

Gene Details

Gene
DYRK3
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004023.1 385 Silent Mutation ACC,ACG T21T NP_001004023.1
NM_003582.2 385 Silent Mutation ACC,ACG T41T NP_003573.2
XM_005273315.4 385 Silent Mutation ACC,ACG T21T XP_005273372.1
XM_011510061.2 385 Silent Mutation ACC,ACG T6T XP_011508363.1

View Full Product Details