Product Details
- SNP ID
-
rs201333896
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:152805220 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCAGAGCTCTGGGGTCTGTGGC[A/G]GTGGGACCTACGGCGCCTGTGGTGG
- Phenotype
-
MIM: 612604
MIM: 612605
MIM: 612606
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LCE1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1048534] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LCE1B
- Gene Name
- late cornified envelope 1B
There are no transcripts associated with this gene.
- Gene
- LCE1C
- Gene Name
- late cornified envelope 1C
- Gene
- LCE1D
- Gene Name
- late cornified envelope 1D
There are no transcripts associated with this gene.
View Full Product Details