Product Details

SNP ID
rs199610627
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:25229020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTTTTTCTTTTCCTCTTCCTTT[A/G]GTTCCCACTCCAAACGAGCTTTTTT
Phenotype
MIM: 607090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYF2 PubMed Links

Gene Details

Gene
SYF2
Gene Name
SYF2 pre-mRNA splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015484.4 291 Missense Mutation CCA,CTA P79L NP_056299.1
NM_207170.3 291 Intron NP_997053.1

View Full Product Details