Product Details

SNP ID
rs199623646
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:10947873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCAGGGTTTGCGTGGATGGAGG[G/T]CTGGACGTGTGTGCAACCCCCCACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf127 PubMed Links

Gene Details

Gene
C1orf127
Gene Name
chromosome 1 open reading frame 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170754.1 2180 Missense Mutation AGA,AGC R754S NP_001164225.1
XM_017000373.1 2180 Missense Mutation AGA,AGC R718S XP_016855862.1

View Full Product Details