Product Details

SNP ID
rs200020097
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:18839875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGTTGGTGGGCAGCTCTTTGCC[A/C]ATGTAGGCGAAGCTGAAACCCACCA
Phenotype
MIM: 606226
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAS1R2 PubMed Links

Gene Details

Gene
TAS1R2
Gene Name
taste 1 receptor member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152232.2 2245 Missense Mutation NP_689418.2

View Full Product Details