Product Details

SNP ID
rs200633484
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:63593490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCCCGGCAGCAAGTCGCCACCA[G/T]GGTGAAGATCGTGACAGTTAAGACC
Phenotype
MIM: 171900
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB7 PubMed Links

Gene Details

Gene
EFCAB7
Gene Name
EF-hand calcium binding domain 7
There are no transcripts associated with this gene.

Gene
PGM1
Gene Name
phosphoglucomutase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172818.1 215 Intron NP_001166289.1
NM_001172819.1 215 Intron NP_001166290.1
NM_002633.2 215 Missense Mutation AGG,ATG R1M NP_002624.2

View Full Product Details