Product Details

SNP ID
rs200142858
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:32895397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTACAGCAAACACGTTCTCCGCCA[A/C]GCAGAGCACTAGGGCGATGCCCCAG
Phenotype
MIM: 142622
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HPCA PubMed Links

Gene Details

Gene
HPCA
Gene Name
hippocalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002143.2 699 Intron NP_002134.2
XM_005270792.2 699 Intron XP_005270849.1
XM_017001118.1 699 Intron XP_016856607.1
Gene
TMEM54
Gene Name
transmembrane protein 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005270408.3 699 Missense Mutation XP_005270465.1

View Full Product Details