Product Details

SNP ID
rs200527448
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:23088939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGGTTGGCCGTAATCGTTCCT[C/T]GGCCCGTACTCGCCCAGACTCTGGC
Phenotype
MIM: 609132 MIM: 601422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM1A PubMed Links

Gene Details

Gene
KDM1A
Gene Name
lysine demethylase 1A
There are no transcripts associated with this gene.

Gene
LUZP1
Gene Name
leucine zipper protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142546.1 3571 Missense Mutation AAG,GAG K1063E NP_001136018.1
NM_033631.3 3571 Missense Mutation AAG,GAG K1063E NP_361013.3
XM_011542090.2 3571 Missense Mutation AAG,GAG K1063E XP_011540392.1
XM_011542091.2 3571 Missense Mutation AAG,GAG K1063E XP_011540393.1
XM_017002252.1 3571 Missense Mutation AAG,GAG K1063E XP_016857741.1

View Full Product Details