Product Details

SNP ID
rs200131751
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:206637753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCTGAACCACCTCCACCCAGA[A/G]GACTAAATGTAAGTAAAAGAAGTCA
Phenotype
MIM: 603497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYRK3 PubMed Links

Gene Details

Gene
DYRK3
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004023.1 443 Missense Mutation AGA,GGA R41G NP_001004023.1
NM_003582.2 443 Missense Mutation AGA,GGA R61G NP_003573.2
XM_005273315.4 443 Missense Mutation AGA,GGA R41G XP_005273372.1
XM_011510061.2 443 Missense Mutation AGA,GGA R26G XP_011508363.1

View Full Product Details