Product Details

SNP ID
rs200476598
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:160371129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTACCTGAACCACGTGCTGGACG[A/T]CTGAACTCAGCCTGTCTCCCACCTC
Phenotype
MIM: 162360
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NHLH1 PubMed Links

Gene Details

Gene
NHLH1
Gene Name
nescient helix-loop-helix 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005598.3 840 Missense Mutation GAC,GTC D133V NP_005589.1

View Full Product Details