Product Details

SNP ID
rs200677081
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:158420431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGCCACAGGCACAGGCAGCAG[A/C]CACTAGTCCCATACACACCCCATGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR10K2 PubMed Links

Gene Details

Gene
OR10K2
Gene Name
olfactory receptor family 10 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004476.1 436 Missense Mutation GCT,TCT A146S NP_001004476.1

View Full Product Details