Product Details

SNP ID
rs200853948
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:39661166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGACGATGCGCTCCGACTCGTC[C/T]GAGAAGAGCACGGCGTCCCCATCGA
Phenotype
MIM: 610525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NT5C1A PubMed Links

Gene Details

Gene
NT5C1A
Gene Name
5'-nucleotidase, cytosolic IA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032526.2 654 Silent Mutation TCA,TCG S218S NP_115915.1

View Full Product Details