Product Details

SNP ID
rs199671355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3778233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTCACCGCTGCTTACGCTCCGC[C/T]TGCTGGAGCCGCCGGGAGAGGTCTT
Phenotype
MIM: 116949 MIM: 117140 MIM: 610674
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf27 PubMed Links

Gene Details

Gene
C20orf27
Gene Name
chromosome 20 open reading frame 27
There are no transcripts associated with this gene.

Gene
CDC25B
Gene Name
cell division cycle 25B
There are no transcripts associated with this gene.

Gene
CENPB
Gene Name
centromere protein B
There are no transcripts associated with this gene.

Gene
SPEF1
Gene Name
sperm flagellar 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015417.4 817 Silent Mutation CAA,CAG Q230Q NP_056232.2
XM_005260683.4 817 UTR 3 XP_005260740.1

View Full Product Details