Product Details

SNP ID
rs200544443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24719780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAATACATGTGGCATAGACAAT[A/G]TGTTAAACGCTGTACTGTATCTGGG
Phenotype
MIM: 610314
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIWIL3 PubMed Links

Gene Details

Gene
PIWIL3
Gene Name
piwi like RNA-mediated gene silencing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008496.3 2921 Missense Mutation CAT,TAT H834Y NP_001008496.2
NM_001255975.1 2921 Missense Mutation CAT,TAT H825Y NP_001242904.1

View Full Product Details